This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Dm02134803_m1
See other CHAT GE Assays ›
Gene Symbol:
ChAT
Gene Name:
Choline acetyltransferase
Gene Aliases:
CG12345, CHAT, CLAT_DROME, CT23399, Cat, ChA, ChAt, Cha, ChaT, DChAT, Dmel\CG12345, Dmel_CG12345, cha, chat, dChAT, l(3)91Cc
Chromosome Location:
Chr.3R: 18705501 - 18732344 on Build Release_6_plus_ISO1_MT
Species:
Drosophila melanogaster
Species Specific ID (Flybase ID):
FBgn0000303
Assay ID Dm02134803_m1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: ChAT
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_057656.3 NP_477004.5 2 - 3 684 57
NM_206517.2 NP_996239.2 2 - 3 684 57
GenBank mRNA M63724.2 - 2 - 3 662 57

Back To Top

More Information


Set Membership:

Amplicon length less than or equal to 70 Recommended Assay Drosophila melanogaster

Panther Classification:

Molecular Function -

acyltransferase

Gene Ontology Categories:

Function(s) Process(es)

neuromuscular synaptic transmission
acetylcholine biosynthetic process
choline O-acetyltransferase activity

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-zpsr4:80/100.66.78.116:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0