This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Dm02137453_g1
See other CRAT GE Assays ›
Gene Symbol:
CRAT
Gene Name:
Carnitine O-Acetyl-Transferase
Gene Aliases:
CG1041, Dmel\CG1041, Dmel_CG1041
Chromosome Location:
Chr.3R: 6371026 - 6376044 on Build Release_6_plus_ISO1_MT
Species:
Drosophila melanogaster
Species Specific ID (Flybase ID):
FBgn0037440
Assay ID Dm02137453_g1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: CRAT
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001043219.1 NP_001036684.1 2 - 3 194 56
NM_001300266.1 NP_001287195.1 2 - 3 178 56
NM_001316490.1 NP_001303419.1 2 - 3 241 56
NM_141393.2 NP_649650.2 2 - 3 291 56
GenBank mRNA BT023787.1 - 2 - 3 137 56

Back To Top

More Information


Set Membership:

Amplicon length less than or equal to 70 Recommended Assay Drosophila melanogaster

Panther Classification:

Molecular Function -

acyltransferase

Gene Ontology Categories:

Function(s) Process(es)

fatty acid beta-oxidation
carnitine metabolic process, CoA-linked
carnitine O-acetyltransferase activity

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-94nls:80/100.66.79.31:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0