This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Dm02137476_g1
See other DMTN GE Assays ›
Gene Symbol:
Dmtn
Gene Name:
Dementin
Gene Aliases:
CG1021, Dmel\CG1021, Dmel_CG1021, anon-WO0118547.375, dmtn
Chromosome Location:
Chr.3R: 6389162 - 6405895 on Build Release_6_plus_ISO1_MT
Species:
Drosophila melanogaster
Species Specific ID (Flybase ID):
FBgn0037443
Assay ID Dm02137476_g1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price 435.00
Your Price
Online offer: 408.65
408.65
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Dmtn
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001144543.3 NP_001138015.1 5 - 6 2703 56
NM_001144544.3 NP_001138016.1 7 - 8 1948 56
NM_001170060.2 NP_001163531.1 6 - 7 1884 56
NM_141397.5 NP_649654.3 6 - 7 2419 56
NM_169141.4 NP_731081.2 6 - 7 2783 56
GenBank mRNA BT011351.1 - 6 - 7 2573 56
BT011461.1 - 5 - 6 2699 56

Back To Top

More Information


Set Membership:

Amplicon length less than or equal to 70 Recommended Assay Drosophila melanogaster

Gene Ontology Categories:

Function(s) Process(es)

brain development
molecular_function

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-mt7zw:80/100.66.79.31:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0