This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Dm02137755_g1
See other EMC1 GE Assays ›
Gene Symbol:
EMC1
Gene Name:
ER membrane protein complex subunit 1
Gene Aliases:
CG2943, Dmel\CG2943, Dmel_CG2943, NEST:bs09f06, emc1
Chromosome Location:
Chr.3R: 7903030 - 7907363 on Build Release_6_plus_ISO1_MT
Species:
Drosophila melanogaster
Species Specific ID (Flybase ID):
FBgn0037530
Assay ID Dm02137755_g1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: EMC1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001300286.1 NP_001287215.1 12 - 13 2727 55
NM_001300287.1 NP_001287216.1 11 - 12 2706 55
NM_001300288.1 NP_001287217.1 11 - 12 2706 55
NM_141490.2 NP_649747.1 12 - 13 2727 55
GenBank mRNA BT031325.1 - 12 - 13 2703 55

Back To Top

More Information


Set Membership:

Amplicon length less than or equal to 70 3' Most Recommended Assay Drosophila melanogaster

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-4x8c5:80/100.66.76.73:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0