This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Dm02138515_g1
See other MPI GE Assays ›
Gene Symbol:
Mpi
Gene Name:
Mannose phosphate isomerase
Gene Aliases:
CG8417, Dmel\CG8417, Dmel_CG8417, MPI, dMPI
Chromosome Location:
Chr.3R: 9759622 - 9761208 on Build Release_6_plus_ISO1_MT
Species:
Drosophila melanogaster
Species Specific ID (Flybase ID):
FBgn0286506
Assay ID Dm02138515_g1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Mpi
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_141683.3 NP_649940.1 2 - 3 734 67
GenBank mRNA AY059444.2 - 2 - 3 722 67
AY075552.1 - 2 - 3 759 67

Back To Top

More Information


Set Membership:

Amplicon length less than or equal to 70 3' Most Recommended Assay Drosophila melanogaster

Panther Classification:

Molecular Function -

isomerase metabolite interconversion enzyme

Gene Ontology Categories:

Function(s) Process(es)

cell wall mannoprotein biosynthetic process
carbohydrate metabolic process
protein glycosylation
GDP-mannose biosynthetic process
mannose-6-phosphate isomerase activity
zinc ion binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-blue-7c59b9b899-ng27p:80/100.66.78.145:80.
git-commit: 8ef7cf686e3c81d28b13492708ce56d167b2f995
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.49.0-Offline