This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Dm02141096_m1
See other PAK3 GE Assays ›
Gene Symbol:
Pak3
Gene Name:
CG14895-PA
Gene Aliases:
CG14895, D-Pak3, DPAK3, DPak3, DmPAK3, Dmel\CG14895, Dmel_CG14895, Dpak3, PAK-3, PAK3, PaK3, anon-WO0118547.285, dPAK3, dpak3, pak3
Chromosome Location:
Chr.3R: 16445007 - 16453821 on Build Release_6_plus_ISO1_MT
Species:
Drosophila melanogaster
Species Specific ID (Flybase ID):
FBgn0044826
Assay ID Dm02141096_m1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Pak3
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001275709.1 NP_001262638.1 1 - 2 2857 70
NM_142288.4 NP_650545.2 1 - 2 2857 70
NM_169719.3 NP_732141.2 1 - 2 2857 70
GenBank mRNA BT003278.1 - 1 - 2 2811 70

Back To Top

More Information


Set Membership:

Amplicon length less than or equal to 70 3' Most Recommended Assay Drosophila melanogaster

Panther Classification:

Molecular Function -

non-receptor serine/threonine protein kinase

Gene Ontology Categories:

Function(s) Process(es)

protein phosphorylation
actin filament organization
myoblast fusion
protein autophosphorylation
positive regulation of synapse assembly
protein serine/threonine kinase activity
ATP binding
protein homodimerization activity
Rac GTPase binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-55658555d6-9rjdb:80/100.66.79.224:80.
git-commit: ec8e7df5f8fec8d765bd419205f1b5046a016d37
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.41.0-Offline