This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Dm02141940_g1
See other VTI1B GE Assays ›
Gene Symbol:
Vti1b
Gene Name:
Vesicle transport through interaction with t-SNAREs 1b
Gene Aliases:
CG31232, CG44009, CG7682, Dmel\CG44009, Dmel_CG44009, EP(3)1037
Chromosome Location:
Chr.3R: 18396995 - 18400339 on Build Release_6_plus_ISO1_MT
Species:
Drosophila melanogaster
Species Specific ID (Flybase ID):
FBgn0264751
Assay ID Dm02141940_g1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Vti1b
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001144603.2 NP_001138075.1 1 - 2 239 57
NM_001275780.1 NP_001262709.1 2 - 3 645 57
GenBank mRNA BT132855.1 - 1 - 2 215 57

Back To Top

More Information


Additional Information:

This assay is in the 5'UTR region of the NM_001275780.1 transcripts.

Set Membership:

Amplicon length less than or equal to 70 3' Most Recommended Assay Drosophila melanogaster

Panther Classification:

Molecular Function -

SNARE protein

Gene Ontology Categories:

Function(s) Process(es)

protein targeting to vacuole
ER to Golgi vesicle-mediated transport
intra-Golgi vesicle-mediated transport
Golgi to vacuole transport
vesicle-mediated transport
retrograde transport, endosome to Golgi
vesicle fusion with Golgi apparatus
SNARE binding
SNAP receptor activity

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-hk65g:80/100.66.77.69:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0