Assay Details
| Gene Symbol: EloA | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_142878.4 | NP_651135.1 | 5 - 6 | 1499 | 52 | |
| NM_170061.3 | NP_732846.1 | 5 - 6 | 1671 | 52 | ||
| GenBank mRNA | AY061363.1 | - | 5 - 6 | 1475 | 52 | |
| BT133197.1 | - | 5 - 6 | 1690 | 52 | ||
More Information
Set Membership: |
|

EloA
Genomic Map