This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Dm02151099_g1
See other ALD1 GE Assays ›
Gene Symbol:
Ald1
Gene Name:
Aldolase 1
Gene Aliases:
ALD, ALDOA, Ald, Aldolase, BcDNA:LP07735, CG6058, Dmel\CG6058, Dmel_CG6058, FBA, T12, T4, T6, T7, ald, dALDOA
Chromosome Location:
Chr.3R: 26254069 - 26261591 on Build Release_6_plus_ISO1_MT
Species:
Drosophila melanogaster
Species Specific ID (Flybase ID):
FBgn0000064
Assay ID Dm02151099_g1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Ald1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_079791.5 NP_524515.2 3 - 4 1406 70
NM_170266.4 NP_733145.3 4 - 5 1395 70
GenBank mRNA BT126163.1 - 3 - 4 1374 70

Back To Top

More Information


Set Membership:

Amplicon length less than or equal to 70 3' Most Drosophila melanogaster

Panther Classification:

Molecular Function -

aldolase

Gene Ontology Categories:

Function(s) Process(es)

glycolytic process
mesoderm development
fructose 1,6-bisphosphate metabolic process
glucose homeostasis
fructose-bisphosphate aldolase activity

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-mt7zw:80/100.66.79.31:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0