Assay Details
| Gene Symbol: slo | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001014651.3 | NP_001014651.1 | 1 - 2 | 767 | 70 | |
| NM_001014652.3 | NP_001014652.1 | 1 - 2 | 767 | 70 | ||
| NM_001014653.3 | NP_001014653.1 | 1 - 2 | 767 | 70 | ||
| NM_001014654.3 | NP_001014654.1 | 1 - 2 | 767 | 70 | ||
| NM_001014655.3 | NP_001014655.1 | 1 - 2 | 767 | 70 | ||
| NM_001014656.3 | NP_001014656.1 | 1 - 2 | 767 | 70 | ||
| NM_001014657.3 | NP_001014657.1 | 1 - 2 | 767 | 70 | ||
| NM_001014658.3 | NP_001014658.1 | 1 - 2 | 767 | 70 | ||
| NM_001014659.3 | NP_001014659.1 | 1 - 2 | 767 | 70 | ||
| NM_001014660.3 | NP_001014660.1 | 1 - 2 | 767 | 70 | ||
| NM_001014662.3 | NP_001014662.1 | 1 - 2 | 767 | 70 | ||
| NM_001014663.3 | NP_001014663.1 | 1 - 2 | 767 | 70 | ||
| NM_001014664.3 | NP_001014664.1 | 1 - 2 | 767 | 70 | ||
| NM_001275996.1 | NP_001262925.1 | 1 - 2 | 767 | 70 | ||
| NM_079762.4 | NP_524486.2 | 1 - 2 | 767 | 70 | ||
More Information
Set Membership: |
|

slo
Genomic Map