This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Dm02374087_s1
See other EST-P GE Assays ›
Gene Symbol:
Est-P thoc6
Gene Name:
Esterase P
CG5632-PA
Gene Aliases:
CG17148, Dmel\CG17148, Dmel_CG17148, EST-5, EST7, ESTP, Est-5, Est-7, Est7, EstP, PsiEst-6, est-P, psiEst-6, yEst-6, yEst6
CG5632, Dmel\CG5632, Dmel_CG5632, THOC6, Thoc6
Chromosome Location:
Chr.3L: 12190733 - 12192528 on Build Release_6_plus_ISO1_MT
Chromosome Location:
Chr.3L: 12190696 - 12193688 on Build Release_6_plus_ISO1_MT
Species:
Drosophila melanogaster
Species Specific ID (Flybase ID):
FBgn0000594
FBgn0036263
Assay ID Dm02374087_s1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Est-P
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_176323.2 NP_788501.1 1 1265 76
GenBank mRNA BT023943.1 - 1 1265 76
Gene Symbol: thoc6
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001316441.1 NP_001303370.1 3 1568 76

Back To Top

More Information


Set Membership:

Amplicon length between 71 and 85 3' Most Drosophila melanogaster

Panther Classification:

Molecular Function -

esterase

Gene Ontology Categories:

Function(s) Process(es)

mRNA export from nucleus in response to heat stress
carboxylic ester hydrolase activity

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-55658555d6-9rjdb:80/100.66.79.224:80.
git-commit: ec8e7df5f8fec8d765bd419205f1b5046a016d37
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.41.0-Offline