This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Dm02607941_g1
See other GLYAT GE Assays ›
Gene Symbol:
Glyat
Gene Name:
Glycine N-acyltransferase
Gene Aliases:
BP1022, CG34010, Dmel\CG34010, Dmel_CG34010, GLYAT, dGLYAT
Chromosome Location:
Chr.2L: 8009603 - 8010652 on Build Release_6_plus_ISO1_MT
Species:
Drosophila melanogaster
Species Specific ID (Flybase ID):
FBgn0054010
Assay ID Dm02607941_g1
Size
Availability Made To Order
Catalog # 4351372
Price 4,430.00
Your Price
Online offer:
4430.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Glyat
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001038794.4 NP_001033883.2 3 - 4 630 103

Back To Top

More Information


Set Membership:

Amplicon length greater than or equal to 101 3' Most Drosophila melanogaster

Gene Ontology Categories:

Function(s) Process(es)

biological_process
molecular_function
transferase activity, transferring acyl groups other than amino-acyl groups

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-zpsr4:80/100.66.78.116:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0