This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Dm02749950_m1
See other MITF GE Assays ›
Gene Symbol:
Mitf
Gene Name:
CG43369-PA
Gene Aliases:
CG17469, CG40476, CG41130, CG43369, Dmel\CG43369, Dmel_CG43369, MITF, MitF, TFEB, dMitf
Chromosome Location:
Chr.4: 1198852 - 1224467 on Build Release_6_plus_ISO1_MT
Species:
Drosophila melanogaster
Species Specific ID (Flybase ID):
FBgn0263112
Assay ID Dm02749950_m1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price 435.00
Your Price
Online offer: 408.65
408.65
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Mitf
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001038718.2 NP_001033807.1 1 - 2 316 79
NM_001038719.2 NP_001033808.1 1 - 2 322 79
NM_001258508.2 NP_001245437.1 1 - 2 316 79
GenBank mRNA AY271259.1 - 1 - 2 273 79
BT011052.1 - 1 - 2 316 79

Back To Top

More Information


Set Membership:

Amplicon length between 71 and 85 3' Most Recommended Assay Drosophila melanogaster

Gene Ontology Categories:

Function(s) Process(es)

compound eye morphogenesis
regulation of transcription, DNA-templated
lysosome organization
cellular response to starvation
lipid catabolic process
positive regulation of transcription, DNA-templated
positive regulation of autophagosome maturation
sequence-specific DNA binding
protein dimerization activity

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-wf7gw:80/100.66.73.223:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0