This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Dm02773276_u1
See other EIF4B GE Assays ›
Gene Symbol:
eIF4B
Gene Name:
eukaryotic translation initiation factor 4B
Gene Aliases:
CG10837, Dm-eIF4B, Dmel\CG10837, Dmel_CG10837, EIF-4B, Eif4B, eIF-4B, eIF-4B-L, eIF-4B-S, eIF-4b, eIF4B-L, eIF4B-S, eIf4b
Chromosome Location:
Chr.3L: 27842669 - 27858271 on Build Release_6_plus_ISO1_MT
Species:
Drosophila melanogaster
Species Specific ID (Flybase ID):
FBgn0020660
Assay ID Dm02773276_u1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: eIF4B
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001015319.2 NP_001015319.1 2 296 102
NM_001015320.2 NP_001015320.1 2 296 102
NM_001015321.2 NP_001015321.1 2 296 102
NM_001276232.1 NP_001263161.1 2 296 102
GenBank mRNA AY071474.1 - 2 302 102
BT015266.1 - 2 259 102

Back To Top

More Information


Set Membership:

Amplicon length greater than or equal to 101 3' Most Recommended Assay Drosophila melanogaster

Panther Classification:

Molecular Function -

translation initiation factor

Gene Ontology Categories:

Function(s) Process(es)

translational initiation
translation initiation factor activity

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-48sk5:80/100.66.77.18:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0