This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Dm03419414_m1
See other RL GE Assays ›
Gene Symbol:
rl
Gene Name:
rolled
Gene Aliases:
12559, BcDNA:RE08694, CG12559, CG18732, CT34260, CT39192, D-ERK, DERK, DERK-A, DmERK, DmERK-A, DmERKA, DmErk, DmMAPK, Dmel\CG12559, Dmel_CG12559, DpERK, DpErk, Dsor2, E(sina)7, EC2-1, EK2-1, ERK, ERK-A, ERK1/2, ERKA, ERKa, EY2-2, Erk, Erk/Map kinase, Erk1, Erk1/2, ErkA, GroupII, MAP-k, MAPK, MapK, Mapk, RL, Rl, SEM, SR2-1, Sem, Su(Raf)2B, dERK, dErk, dm-dpERK, dp-ERK, dpERK, dpERK1, dpERk, dpErk, dpMAPK, erk, l(2)41Ac, l(2R)EMS45-39, mapk, p-ERK, pERK, pMAPK, pMapK, rl/ERK, rl/MAPK, rll, sem
Chromosome Location:
Chr.2R: 1071462 - 1125927 on Build Release_6_plus_ISO1_MT
Species:
Drosophila melanogaster
Species Specific ID (Flybase ID):
FBgn0003256
Assay ID Dm03419414_m1
Size
Availability Made To Order
Catalog # 4351372
Price 435.00
Your Price
Online offer: 408.65
408.65
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: rl
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001015121.3 NP_001015121.3 5 - 6 587 147
NM_001015122.3 NP_001015122.1 4 - 5 518 147
NM_001015123.3 NP_001015123.1 3 - 4 699 147
NM_001110878.4 NP_001104348.2 5 - 6 556 147
NM_001110879.2 NP_001104349.1 4 - 5 477 147
NM_001300706.2 NP_001287635.1 4 - 5 494 147
GenBank mRNA AY070996.1 - 4 - 5 518 147
M95124.1 - 3 - 4 644 147

Back To Top

More Information


Additional Information:

This assay is in the 5'UTR region of the NM_001110878.4 transcripts.

Set Membership:

Amplicon length greater than or equal to 101 Drosophila melanogaster

Panther Classification:

Molecular Function -

non-receptor serine/threonine protein kinase

Gene Ontology Categories:

Function(s) Process(es)

MAPK cascade
mitotic cell cycle
cell surface receptor signaling pathway
epidermal growth factor receptor signaling pathway
terminal region determination
terminal branching, open tracheal system
R7 cell fate commitment
imaginal disc-derived wing vein specification
imaginal disc-derived wing morphogenesis
leg disc proximal/distal pattern formation
metamorphosis
positive regulation of cell proliferation
insulin receptor signaling pathway
torso signaling pathway
determination of adult lifespan
fibroblast growth factor receptor signaling pathway
cellular response to starvation
regulation of gene expression
negative regulation of macroautophagy
cellular response to reactive oxygen species
lymph gland plasmatocyte differentiation
lymph gland crystal cell differentiation
tracheal pit formation in open tracheal system
intracellular signal transduction
sevenless signaling pathway
positive regulation of cell size
positive regulation of photoreceptor cell differentiation
dorsal appendage formation
vascular endothelial growth factor receptor signaling pathway
behavioral response to ethanol
modulation of synaptic transmission
positive regulation of sequence-specific DNA binding transcription factor activity
defense response to virus
branching involved in open tracheal system development
ERK1 and ERK2 cascade
cellular response to arsenic-containing substance
cellular response to cadmium ion
positive regulation of wound healing
regulation of heart morphogenesis
regulation of response to drug
protein serine/threonine kinase activity
MAP kinase activity
ATP binding
transcription factor binding
protein kinase binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-6b469b8bb7-msbq7:80/100.66.78.64:80.
git-commit: a334af76dff23450325448aedefe62379591458a
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.48.1-2026.04.16-1.0