Assay Details
| Gene Symbol: lt | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001042950.3 | NP_001036415.2 | 11 - 12 | 2250 | 83 | |
| NM_001042951.2 | NP_001036416.1 | 11 - 12 | 2250 | 83 | ||
| NM_001042952.2 | NP_001036417.1 | 10 - 11 | 2302 | 83 | ||
| NM_001273766.1 | NP_001260695.1 | 11 - 12 | 2250 | 83 | ||
| GenBank mRNA | AY118388.1 | - | 10 - 11 | 2310 | 83 | |
| BT024228.1 | - | 11 - 12 | 2207 | 83 | ||
More Information
Set Membership: |
|

lt
Genomic Map