This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Dm03421070_m1
See other SNAP25 GE Assays ›
Gene Symbol:
Snap25
Gene Name:
Synaptosomal-associated protein 25kDa
Gene Aliases:
CG17676, CG17884, CG40452, Dmel\CG40452, Dmel_CG40452, SNAP-25, SNAP25, dSNAP-25, snap-25, snap25, snp25
Chromosome Location:
Chr.3L: 24074180 - 24299205 on Build Release_6_plus_ISO1_MT
Species:
Drosophila melanogaster
Species Specific ID (Flybase ID):
FBgn0011288
Assay ID Dm03421070_m1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Snap25
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001043176.2 NP_001036641.1 2 - 3 226 71
NM_001043177.3 NP_001036642.2 2 - 3 226 71
GenBank mRNA BT023789.1 - 2 - 3 177 71
BT126217.1 - 2 - 3 178 71
L22021.1 - 2 - 3 175 71

Back To Top

More Information


Set Membership:

Amplicon length between 71 and 85 Recommended Assay Drosophila melanogaster

Panther Classification:

Molecular Function -

SNARE protein

Gene Ontology Categories:

Function(s) Process(es)

exocytosis
Golgi to plasma membrane transport
vesicle fusion
neurotransmitter secretion
neuromuscular synaptic transmission
synaptic vesicle docking
synaptic vesicle priming
vesicle-mediated transport
synaptic vesicle fusion to presynaptic active zone membrane
regulation of short-term neuronal synaptic plasticity
synaptic vesicle transport
SNARE binding
SNAP receptor activity
protein binding
syntaxin binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-55658555d6-pnd5g:80/100.66.79.83:80.
git-commit: ec8e7df5f8fec8d765bd419205f1b5046a016d37
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.41.0-Offline