This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Dr03071155_m1
See other ECI2 GE Assays ›
Gene Symbol:
eci2
Gene Name:
enoyl-CoA delta isomerase 2
Gene Aliases:
PECI, fd61c12, wu:fd61c12, zgc:175023, zgc:92030
Chromosome Location:
Chr.24: 2956352 - 2972040 on Build GRCz11
Species:
Zebrafish
Species Specific ID (Flybase ID):
-
Assay ID Dr03071155_m1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: eci2
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001002645.3 NP_001002645.3 9 - 10 1139 58
XM_021470136.1 XP_021325811.1 9 - 10 1050 58
XM_021470137.1 XP_021325812.1 9 - 10 1129 58
GenBank mRNA BC076531.1 - 9 - 10 1066 58
EH523368.1 - 1 - 2 643 58

Back To Top

More Information


Set Membership:

Amplicon length less than or equal to 70 3' Most Mammalian Gene Collection Recommended Assay Danio rerio Probe spans exons

Panther Classification:

Molecular Function -

transfer/carrier protein

Gene Ontology Categories:

Process(es)

fatty-acyl-CoA binding
catalytic activity

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-blue-7c59b9b899-9dqhd:80/100.66.72.70:80.
git-commit: 8ef7cf686e3c81d28b13492708ce56d167b2f995
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.49.0-Offline