This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Dr03072818_m1
See other CADM3 GE Assays ›
Gene Symbol:
cadm3
Gene Name:
cell adhesion molecule 3
Gene Aliases:
zgc:136589
Chromosome Location:
Chr.2: 2926 - 93004 on Build GRCz11
Species:
Zebrafish
Species Specific ID (Flybase ID):
-
Assay ID Dr03072818_m1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price 4,430.00
Your Price
Online offer:
4430.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: cadm3
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001045246.1 NP_001038711.1 7 - 8 957 69
XM_005171391.4 XP_005171448.1 7 - 8 1252 69
XM_009298778.3 XP_009297053.1 7 - 8 1252 69
XM_021481265.1 XP_021336940.1 7 - 8 1252 69
GenBank mRNA BC116600.1 - 7 - 8 957 69
CU638752.1 - 7 - 8 1104 69
EU182354.1 - 6 - 7 825 69

Back To Top

More Information


Set Membership:

Amplicon length less than or equal to 70 3' Most Mammalian Gene Collection Recommended Assay Danio rerio Probe spans exons

Gene Ontology Categories:

Process(es)

protein binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-zpsr4:80/100.66.78.116:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0