This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Dr03072837_m1
See other CDH4 GE Assays ›
Gene Symbol:
cdh4
Gene Name:
cadherin 4, type 1, R-cadherin (retinal)
Gene Aliases:
R-cadherin, rcad, zgc:136316
Chromosome Location:
Chr.11: 49754 - 53395 on Build GRCz11
Species:
Zebrafish
Species Specific ID (Flybase ID):
-
Assay ID Dr03072837_m1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price 435.00
Your Price
Online offer: 408.65
408.65
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: cdh4
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001045273.2 NP_001038738.2 1 - 2 218 67
NM_131086.2 NP_571161.2 1 - 2 162 67
GenBank mRNA BC116496.1 - 1 - 2 165 67
BC129136.1 - 1 - 2 162 67
BC135069.1 - 1 - 2 218 67

Back To Top

More Information


Set Membership:

Amplicon length less than or equal to 70 3' Most Mammalian Gene Collection Recommended Assay Danio rerio Probe spans exons

Panther Classification:

Molecular Function -

cadherin

Gene Ontology Categories:

Function(s) Process(es)

cell adhesion
homophilic cell adhesion via plasma membrane adhesion molecules
cranial nerve development
regulation of eye photoreceptor cell development
camera-type eye morphogenesis
lateral line system development
retina morphogenesis in camera-type eye
cell-cell adhesion
cell-cell adhesion via plasma-membrane adhesion molecules
calcium ion binding
cadherin binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-mt7zw:80/100.66.79.31:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0