This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Dr03072878_m1
See other CYB5R2 GE Assays ›
Gene Symbol:
cyb5r2
Gene Name:
cytochrome b5 reductase 2
Gene Aliases:
im:7147295, zgc:153291
Chromosome Location:
Chr.25: 16046615 - 16057540 on Build GRCz11
Species:
Zebrafish
Species Specific ID (Flybase ID):
-
Assay ID Dr03072878_m1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price 435.00
Your Price
Online offer: 408.65
408.65
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: cyb5r2
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001045360.1 NP_001038825.1 2 - 3 254 78
GenBank mRNA BC122221.1 - 2 - 3 254 78

Back To Top

More Information


Set Membership:

Amplicon length between 71 and 85 Mammalian Gene Collection Recommended Assay Danio rerio Probe spans exons

Gene Ontology Categories:

Function(s) Process(es)

response to yeast
lipid metabolic process
steroid biosynthetic process
steroid metabolic process
sterol biosynthetic process
oxidation-reduction process
cytochrome-b5 reductase activity, acting on NAD(P)H
oxidoreductase activity
FAD binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-nwsk8:80/100.66.76.192:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0