Assay Details
| Gene Symbol: ldb3b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_199858.3 | NP_956152.1 | 2 - 3 | 382 | 70 | |
| XM_005156271.4 | XP_005156328.1 | 3 - 4 | 431 | 70 | ||
| XM_005156272.4 | XP_005156329.1 | 3 - 4 | 436 | 70 | ||
| XM_005156273.4 | XP_005156330.1 | 3 - 4 | 482 | 70 | ||
| XM_009306591.3 | XP_009304866.1 | 3 - 4 | 477 | 70 | ||
| GenBank mRNA | BC047189.1 | - | 3 - 4 | 417 | 70 | |
| BC065846.1 | - | 2 - 3 | 387 | 70 | ||
More Information
Set Membership: |
|

ldb3b
Genomic Map