This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Dr03074671_g1
See other HACD2 GE Assays ›
Gene Symbol:
hacd2
Gene Name:
3-hydroxyacyl-CoA dehydratase 2
Gene Aliases:
ptplb, wu:fi32c08, zgc:55421
Chromosome Location:
Chr.9: 38029475 - 38042889 on Build GRCz11
Species:
Zebrafish
Species Specific ID (Flybase ID):
-
Assay ID Dr03074671_g1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: hacd2
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_199861.1 NP_956155.1 5 - 6 527 68
GenBank mRNA BC044396.1 - 5 - 6 527 68
BC165084.1 - 5 - 6 489 68

Back To Top

More Information


Set Membership:

Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Danio rerio Probe spans exons

Gene Ontology Categories:

Function(s) Process(es)

lipid metabolic process
fatty acid metabolic process
fatty acid biosynthetic process
sphingolipid biosynthetic process
fatty acid elongation
very long-chain fatty acid biosynthetic process
lyase activity
3-hydroxyacyl-CoA dehydratase activity
very-long-chain 3-hydroxyacyl-CoA dehydratase activity
3-hydroxy-arachidoyl-CoA dehydratase activity
3-hydroxy-behenoyl-CoA dehydratase activity
3-hydroxy-lignoceroyl-CoA dehydratase activity

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-jbr9k:80/100.66.74.50:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0