This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Dr03074798_m1
See other HAGH GE Assays ›
Gene Symbol:
hagh
Gene Name:
hydroxyacylglutathione hydrolase
Gene Aliases:
fa66a03, wu:fa66a03, zgc:73161
Chromosome Location:
Chr.3: 18774509 - 18792570 on Build GRCz11
Species:
Zebrafish
Species Specific ID (Flybase ID):
-
Assay ID Dr03074798_m1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: hagh
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_200043.2 NP_956337.1 2 - 3 255 67
XM_005163865.4 XP_005163922.1 2 - 3 319 67
XM_009299331.3 XP_009297606.1 2 - 3 319 67
XM_021469713.1 XP_021325388.1 2 - 3 319 67
GenBank mRNA AY398356.1 - 2 - 3 255 67
BC060913.1 - 1 - 2 143 67
BC066607.1 - 2 - 3 241 67

Back To Top

More Information


Set Membership:

Amplicon length less than or equal to 70 Mammalian Gene Collection Recommended Assay Danio rerio Probe spans exons

Gene Ontology Categories:

Function(s) Process(es)

methylglyoxal catabolic process to D-lactate via S-lactoyl-glutathione
hydroxyacylglutathione hydrolase activity
hydrolase activity
metal ion binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-zpsr4:80/100.66.78.116:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0