This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Dr03075674_m1
See other MYD88 GE Assays ›
Gene Symbol:
myd88
Gene Name:
MYD88 innate immune signal transduction adaptor
Gene Aliases:
zgc:103541
Chromosome Location:
Chr.24: 20188914 - 20192854 on Build GRCz11
Species:
Zebrafish
Species Specific ID (Flybase ID):
-
Assay ID Dr03075674_m1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: myd88
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_212814.2 NP_997979.2 4 - 5 831 84
GenBank mRNA AY388401.1 - 4 - 5 695 84
AY389463.1 - 2 - 3 276 84
BC083419.1 - 4 - 5 831 84
BC164642.1 - 4 - 5 729 84
DQ100359.1 - 4 - 5 751 84

Back To Top

More Information


Set Membership:

Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Recommended Assay Danio rerio Probe spans exons

Panther Classification:

Molecular Function -

scaffold/adaptor protein

Gene Ontology Categories:

Function(s) Process(es)

activation of innate immune response
response to molecule of bacterial origin
neutrophil activation involved in immune response
immune system process
MyD88-dependent toll-like receptor signaling pathway
inflammatory response
signal transduction
Toll signaling pathway
neutrophil chemotaxis
monocyte activation
defense response to bacterium
positive regulation of I-kappaB kinase/NF-kappaB signaling
innate immune response
defense response to Gram-positive bacterium
regulation of hematopoietic progenitor cell differentiation
positive regulation of hematopoietic progenitor cell differentiation
protein binding
Toll-like receptor binding
TIR domain binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-rpjph:80/100.66.75.107:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0