This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Dr03077033_m1
See other PLEKHA8 GE Assays ›
Gene Symbol:
plekha8
Gene Name:
pleckstrin homology domain containing, family A (phosphoinositide binding specific) member 8
Gene Aliases:
fc68b04, wu:fc68b04, zgc:101886
Chromosome Location:
Chr.16: 20267814 - 20294234 on Build GRCz11
Species:
Zebrafish
Species Specific ID (Flybase ID):
-
Assay ID Dr03077033_m1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price 435.00
Your Price
Online offer: 330.60
330.6
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: plekha8
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001007374.1 NP_001007375.1 4 - 5 534 66
GenBank mRNA BC085465.1 - 4 - 5 534 66

Back To Top

More Information


Set Membership:

Amplicon length less than or equal to 70 Mammalian Gene Collection Recommended Assay Danio rerio Probe spans exons

Panther Classification:

Molecular Function -

transfer/carrier protein

Gene Ontology Categories:

Function(s) Process(es)

protein transport
ER to Golgi ceramide transport
ceramide transport
intermembrane lipid transfer
lipid binding
intermembrane lipid transfer activity
ceramide 1-phosphate binding
ceramide 1-phosphate transporter activity

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-6b469b8bb7-p58g2:80/100.66.72.198:80.
git-commit: a334af76dff23450325448aedefe62379591458a
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.48.1-2026.04.16-1.0