This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Dr03078718_g1
See other BATF3 GE Assays ›
Gene Symbol:
batf3
Gene Name:
basic leucine zipper transcription factor, ATF-like 3
Gene Aliases:
si:ch211-147d7.2, zgc:153485
Chromosome Location:
Chr.20: 37812699 - 37813667 on Build GRCz11
Species:
Zebrafish
Species Specific ID (Flybase ID):
-
Assay ID Dr03078718_g1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: batf3
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001045392.1 NP_001038857.1 1 - 2 77 86
GenBank mRNA BC122303.1 - 1 - 2 77 86
BC164923.1 - 1 - 2 108 86

Back To Top

More Information


Set Membership:

Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Danio rerio Probe spans exons

Gene Ontology Categories:

Function(s) Process(es)

regulation of transcription, DNA-templated
regulation of transcription from RNA polymerase II promoter
RNA polymerase II core promoter proximal region sequence-specific DNA binding
RNA polymerase II transcription factor activity, sequence-specific DNA binding
DNA binding
transcription factor activity, sequence-specific DNA binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-nwsk8:80/100.66.76.192:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0