Assay Details
| Gene Symbol: foxr1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001103124.1 | NP_001096594.1 | 5 - 6 | 983 | 101 | |
| GenBank mRNA | BC151951.1 | - | 5 - 6 | 983 | 101 | |
More Information
Set Membership: |
|

foxr1
Genomic Map