This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Dr03081321_m1
See other EIF6 GE Assays ›
Gene Symbol:
eif6
Gene Name:
eukaryotic translation initiation factor 6
Gene Aliases:
Itgb4bp, eIF-6, itgb4bp4, wu:fc28a04, wu:ft88f05, zgc:56562
Chromosome Location:
Chr.6: 52339637 - 52348629 on Build GRCz11
Species:
Zebrafish
Species Specific ID (Flybase ID):
-
Assay ID Dr03081321_m1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price 4,430.00
Your Price
Online offer:
4430.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: eif6
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_200944.1 NP_957238.1 4 - 5 448 77
GenBank mRNA AY398366.1 - 4 - 5 459 77
BC049488.1 - 4 - 5 448 77
BC071374.1 - 4 - 5 460 77

Back To Top

More Information


Set Membership:

Amplicon length between 71 and 85 Mammalian Gene Collection Recommended Assay Danio rerio Probe spans exons

Gene Ontology Categories:

Function(s) Process(es)

ribosomal subunit export from nucleus
maturation of 5.8S rRNA
maturation of LSU-rRNA
translation
translational initiation
ribosome biogenesis
mature ribosome assembly
assembly of large subunit precursor of preribosome
translation initiation factor activity
ribosome binding
ribosomal large subunit binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-zpsr4:80/100.66.78.116:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0