This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Dr03081385_m1
See other PPIL2 GE Assays ›
Gene Symbol:
ppil2
Gene Name:
peptidylprolyl isomerase (cyclophilin)-like 2
Gene Aliases:
si:dkeyp-86g2.1, zgc:56616, zgc:86735
Chromosome Location:
Chr.5: 67234 - 138428 on Build GRCz11
Species:
Zebrafish
Species Specific ID (Flybase ID):
-
Assay ID Dr03081385_m1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: ppil2
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_200991.2 NP_957285.2 3 - 4 237 72
GenBank mRNA BC049506.1 - 3 - 4 189 72
BC071410.1 - 3 - 4 237 72

Back To Top

More Information


Set Membership:

Amplicon length between 71 and 85 Mammalian Gene Collection Recommended Assay Danio rerio Probe spans exons

Panther Classification:

Molecular Function -

chaperone

Gene Ontology Categories:

Function(s) Process(es)

protein polyubiquitination
protein peptidyl-prolyl isomerization
protein folding
protein ubiquitination
peptidyl-prolyl cis-trans isomerase activity
ubiquitin-protein transferase activity
isomerase activity
ubiquitin-ubiquitin ligase activity

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-fxmhk:80/100.66.79.98:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0