This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Dr03082260_m1
See other NAA10 GE Assays ›
Gene Symbol:
naa10
Gene Name:
N-alpha-acetyltransferase 10, NatA catalytic subuni
Gene Aliases:
ard1a, hm:zeh0223, wu:fc66b08, zgc:63981
Chromosome Location:
Chr.23: 13832347 - 13840404 on Build GRCz11
Species:
Zebrafish
Species Specific ID (Flybase ID):
-
Assay ID Dr03082260_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: naa10
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_213334.1 NP_998499.1 6 - 7 505 132
GenBank mRNA BC053180.1 - 6 - 7 505 132
BC165054.1 - 6 - 7 421 132

Back To Top

More Information


Set Membership:

Amplicon length greater than or equal to 101 Mammalian Gene Collection Danio rerio Probe spans exons

Panther Classification:

Molecular Function -

acetyltransferase

Gene Ontology Categories:

Function(s) Process(es)

N-terminal peptidyl-serine acetylation
N-terminal peptidyl-glutamic acid acetylation
peptide alpha-N-acetyltransferase activity
N-acetyltransferase activity
transferase activity
peptide-serine-N-acetyltransferase activity
peptide-glutamate-N-acetyltransferase activity

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-9hc65:80/100.66.78.230:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0