Assay Details
| Gene Symbol: tspan5b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001008591.1 | NP_001008591.1 | 4 - 5 | 720 | 85 | |
| GenBank mRNA | BC086749.1 | - | 4 - 5 | 720 | 85 | |
| BC164841.1 | - | 4 - 5 | 484 | 85 | ||
More Information
Set Membership: |
|

tspan5b
Genomic Map