This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Dr03084831_m1
See other TNFRSF19 GE Assays ›
Gene Symbol:
tnfrsf19
Gene Name:
tumor necrosis factor receptor superfamily, member 19
Gene Aliases:
fa04d10, si:ch211-113p18.2, troy, wu:fa04d10, wu:fk80e07, zgc:136805
Chromosome Location:
Chr.15: 8235531 - 8309243 on Build GRCz11
Species:
Zebrafish
Species Specific ID (Flybase ID):
-
Assay ID Dr03084831_m1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price 435.00
Your Price
Online offer: 408.65
408.65
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: tnfrsf19
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001044904.1 NP_001038369.1 3 - 4 392 58
XM_017350963.2 XP_017206452.1 3 - 4 518 58
XM_021481422.1 XP_021337097.1 3 - 4 584 58
XM_021481423.1 XP_021337098.1 2 - 3 442 58
GenBank mRNA BC117651.1 - 3 - 4 392 58
EF426728.1 - 3 - 4 431 58

Back To Top

More Information


Set Membership:

Amplicon length less than or equal to 70 Mammalian Gene Collection Recommended Assay Danio rerio Probe spans exons

Gene Ontology Categories:

Function(s) Process(es)

sprouting angiogenesis
establishment of endothelial blood-brain barrier
positive regulation of I-kappaB kinase/NF-kappaB signaling
positive regulation of JNK cascade
posterior lateral line development
posterior lateral line neuromast deposition
protein binding
signaling receptor activity

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-l4hk6:80/100.66.76.55:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0