This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Dr03086055_m1
See other GPAT2 GE Assays ›
Gene Symbol:
gpat2
Gene Name:
glycerol-3-phosphate acyltransferase 2, mitochondrial
Gene Aliases:
im:7149526, zgc:162485
Chromosome Location:
Chr.8: 41035230 - 41173369 on Build GRCz11
Species:
Zebrafish
Species Specific ID (Flybase ID):
-
Assay ID Dr03086055_m1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price 4,430.00
Your Price
Online offer:
4430.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: gpat2
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001089380.1 NP_001082849.1 9 - 10 796 60
GenBank mRNA BC093346.1 - 8 - 9 700 60
BC139593.1 - 9 - 10 796 60

Back To Top

More Information


Set Membership:

Amplicon length less than or equal to 70 Mammalian Gene Collection Recommended Assay Danio rerio Probe spans exons

Gene Ontology Categories:

Function(s) Process(es)

glycerol-3-phosphate metabolic process
fatty acid metabolic process
phospholipid biosynthetic process
triglyceride biosynthetic process
cellular lipid metabolic process
piRNA biosynthetic process
glycerol-3-phosphate O-acyltransferase activity
O-acyltransferase activity

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-zpsr4:80/100.66.78.116:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0