This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Dr03088180_m1
See other CBLL1 GE Assays ›
Gene Symbol:
cbll1
Gene Name:
Cbl proto-oncogene-like 1, E3 ubiquitin protein ligase
Gene Aliases:
zgc:55308
Chromosome Location:
Chr.4: 18836791 - 18841071 on Build GRCz11
Species:
Zebrafish
Species Specific ID (Flybase ID):
-
Assay ID Dr03088180_m1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price 435.00
Your Price
Online offer: 408.65
408.65
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: cbll1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001309368.1 NP_001296297.1 3 - 4 372 68
NM_001309369.1 NP_001296298.1 3 - 4 332 68
NM_201066.2 NP_957360.2 3 - 4 311 68
XM_017354982.2 XP_017210471.1 3 - 4 355 68
GenBank mRNA BC048028.1 - 3 - 4 311 68

Back To Top

More Information


Additional Information:

This assay is in the 5'UTR region of the NM_001309369.1 transcripts.

Set Membership:

Amplicon length less than or equal to 70 Mammalian Gene Collection Recommended Assay Danio rerio Probe spans exons

Gene Ontology Categories:

Function(s) Process(es)

protein ubiquitination
regulation of cell adhesion
ubiquitin protein ligase activity

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-mt7zw:80/100.66.79.31:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0