This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Dr03090486_m1
See other STARD4 GE Assays ›
Gene Symbol:
stard4
Gene Name:
StAR-related lipid transfer (START) domain containing 4
Gene Aliases:
zgc:112466
Chromosome Location:
Chr.21: 244466 - 258069 on Build GRCz11
Species:
Zebrafish
Species Specific ID (Flybase ID):
-
Assay ID Dr03090486_m1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price 4,430.00
Your Price
Online offer:
4430.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: stard4
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001017679.1 NP_001017679.2 4 - 5 297 78
XM_005170234.3 XP_005170291.1 4 - 5 373 78
XM_017352776.2 XP_017208265.1 3 - 4 304 78
XM_021468820.1 XP_021324495.1 5 - 6 387 78
GenBank mRNA BC093326.1 - 4 - 5 297 78

Back To Top

More Information


Set Membership:

Amplicon length between 71 and 85 Mammalian Gene Collection Recommended Assay Danio rerio Probe spans exons

Gene Ontology Categories:

Function(s) Process(es)

cholesterol transport involved in cholesterol storage
intracellular cholesterol transport
cholesterol import
lipid binding
cholesterol binding
intermembrane cholesterol transfer activity

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-blue-7c59b9b899-49tpj:80/100.66.77.222:80.
git-commit: 8ef7cf686e3c81d28b13492708ce56d167b2f995
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.49.0-Offline