This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Dr03091347_mH
See other FZD3A GE Assays ›
Gene Symbol:
fzd3a
Gene Name:
frizzled class receptor 3a
Gene Aliases:
fz9, fzd3, zg09
Chromosome Location:
Chr.20: 35208851 - 35246212 on Build GRCz11
Species:
Zebrafish
Species Specific ID (Flybase ID):
-
Assay ID Dr03091347_mH
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: fzd3a
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001042761.2 NP_001036226.1 2 - 3 674 92
XM_005160748.4 XP_005160805.1 2 - 3 726 92
GenBank mRNA AB246776.1 - 1 - 2 199 92
BC161660.1 - 2 - 3 658 92
BC163439.1 - 1 - 2 329 92
BC163452.1 - 1 - 2 329 92
DQ200953.1 - 2 - 3 674 92

Back To Top

More Information


Set Membership:

Amplicon length between 86 and 100 G-protein coupled receptor Mammalian Gene Collection Danio rerio Probe spans exons

Panther Classification:

Molecular Function -

transmembrane signal receptor

Gene Ontology Categories:

Function(s) Process(es)

cell surface receptor signaling pathway
multicellular organism development
Wnt signaling pathway
facial nucleus development
anterior/posterior axon guidance
non-canonical Wnt signaling pathway
canonical Wnt signaling pathway
commissural neuron axon guidance
transmembrane signaling receptor activity
Wnt-protein binding
Wnt-activated receptor activity

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-tks2p:80/100.66.79.31:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0