This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Dr03092354_g1
See other SKP2 GE Assays ›
Gene Symbol:
skp2
Gene Name:
S-phase kinase-associated protein 2, E3 ubiquitin protein ligase
Gene Aliases:
id:ibd5121, sb:cb933, wu:fa10b08, wu:fj97b11, zgc:162950
Chromosome Location:
Chr.21: 22347120 - 22357987 on Build GRCz11
Species:
Zebrafish
Species Specific ID (Flybase ID):
-
Assay ID Dr03092354_g1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price 435.00
Your Price
Online offer: 408.65
408.65
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: skp2
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001082837.2 NP_001076306.1 8 - 9 1074 98
XM_005161189.4 XP_005161246.1 8 - 9 977 98
GenBank mRNA BC134165.1 - 8 - 9 1038 98

Back To Top

More Information


Set Membership:

Amplicon length between 86 and 100 3' Most Mammalian Gene Collection Detect Genomic DNA Recommended Assay Danio rerio Probe spans exons

Gene Ontology Categories:

Function(s)

G2/M transition of mitotic cell cycle
SCF-dependent proteasomal ubiquitin-dependent protein catabolic process
regulation of cell cycle

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-mt7zw:80/100.66.79.31:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0