This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Dr03094120_m1
See other RPL10 GE Assays ›
Gene Symbol:
rpl10
Gene Name:
ribosomal protein L10
Gene Aliases:
QM, QM protein, fa93d03, rpl-10, wu:fa93d03, zgc:56154
Chromosome Location:
Chr.23: 19712556 - 19715814 on Build GRCz11
Species:
Zebrafish
Species Specific ID (Flybase ID):
-
Assay ID Dr03094120_m1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: rpl10
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_200027.1 NP_956321.1 3 - 4 218 83
XM_017353523.1 XP_017209012.1 4 - 5 232 83
GenBank mRNA AY763500.1 - 3 - 4 220 83
BC045950.1 - 3 - 4 218 83
BC164313.1 - 3 - 4 229 83

Back To Top

More Information


Set Membership:

Amplicon length between 71 and 85 Mammalian Gene Collection Recommended Assay Danio rerio Probe spans exons

Panther Classification:

Molecular Function -

ribosomal protein

Gene Ontology Categories:

Function(s) Process(es)

ribosomal large subunit assembly
translation
regulation of translation
multicellular organism development
brain development
embryonic brain development
structural constituent of ribosome
translation regulator activity

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-blue-7c59b9b899-zvjvc:80/100.66.72.70:80.
git-commit: 8ef7cf686e3c81d28b13492708ce56d167b2f995
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.49.0-Offline