This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Dr03094798_m1
See other EIF4H GE Assays ›
Gene Symbol:
eif4h
Gene Name:
eukaryotic translation initiation factor 4h
Gene Aliases:
wbscr1, zgc:77282
Chromosome Location:
Chr.15: 4585401 - 4616819 on Build GRCz11
Species:
Zebrafish
Species Specific ID (Flybase ID):
-
Assay ID Dr03094798_m1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: eif4h
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001197328.1 NP_001184257.1 7 - 8 852 69
NM_001197329.1 NP_001184258.1 6 - 7 687 69
NM_205695.2 NP_991258.2 7 - 8 834 69
XM_009291449.3 XP_009289724.1 6 - 7 765 69
GenBank mRNA BC056285.1 - 7 - 8 779 69
BC065640.1 - 7 - 8 800 69
CT633866.2 - 6 - 7 613 69
EH515860.1 - 1 - 2 508 69

Back To Top

More Information


Set Membership:

Amplicon length less than or equal to 70 3' Most Mammalian Gene Collection Recommended Assay Danio rerio Probe spans exons

Panther Classification:

Molecular Function -

translation initiation factor

Gene Ontology Categories:

Function(s) Process(es)

formation of translation preinitiation complex
cytoplasmic translation
eukaryotic translation initiation factor 4F complex assembly
nucleic acid binding
RNA binding
RNA strand annealing activity
RNA strand-exchange activity
ribosomal small subunit binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-jbr9k:80/100.66.74.50:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0