This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Dr03094809_m1
See other UNC119B GE Assays ›
Gene Symbol:
unc119b
Gene Name:
unc-119 homolog b (C. elegans)
Gene Aliases:
Unc119, unc119.2, zfUnc119a, zgc:136476
Chromosome Location:
Chr.21: 39121453 - 39177430 on Build GRCz11
Species:
Zebrafish
Species Specific ID (Flybase ID):
-
Assay ID Dr03094809_m1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: unc119b
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001040394.2 NP_001035484.1 2 - 3 422 69
GenBank mRNA AF387341.1 - 2 - 3 261 69
BC074025.1 - 2 - 3 347 69
BC094960.1 - 2 - 3 336 69
BC115147.1 - 2 - 3 556 69
BC124479.1 - 2 - 3 422 69
BC129251.1 - 2 - 3 535 69
BC164428.1 - 2 - 3 446 69

Back To Top

More Information


Set Membership:

Amplicon length less than or equal to 70 Mammalian Gene Collection Recommended Assay Danio rerio Probe spans exons

Panther Classification:

Molecular Function -

membrane traffic protein

Gene Ontology Categories:

Function(s) Process(es)

mitotic cytokinesis
nervous system development
visual perception
lipoprotein transport
positive regulation of protein tyrosine kinase activity
Kupffer's vesicle development
negative regulation of clathrin-mediated endocytosis
negative regulation of caveolin-mediated endocytosis
lipid binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-rr9bx:80/100.66.76.204:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0