This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Dr03095013_m1
See other DND1 GE Assays ›
Gene Symbol:
dnd1
Gene Name:
DND microRNA-mediated repression inhibitor 1
Gene Aliases:
cb736, dnd, wu:fi22d10, zgc:111864
Chromosome Location:
Chr.14: 7397889 - 7409019 on Build GRCz11
Species:
Zebrafish
Species Specific ID (Flybase ID):
-
Assay ID Dr03095013_m1
Best Coverage
Size
Availability Inventoried
Catalog # 4331182
Price 284.00
Your Price
Online offer: 266.65
266.65
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: dnd1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_212795.1 NP_997960.1 3 - 4 670 80
GenBank mRNA AY225448.1 - 3 - 4 670 80
BC095578.1 - 3 - 4 632 80
BC164513.1 - 3 - 4 641 80

Back To Top

More Information


Set Membership:

Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Recommended Assay Danio rerio Inventoried Probe spans exons

Panther Classification:

Molecular Function -

RNA metabolism protein

Gene Ontology Categories:

Function(s) Process(es)

multicellular organism development
germ cell development
germ cell migration
positive regulation of gene expression
mRNA stabilization
heart contraction
negative regulation of gene silencing by miRNA
regulation of cell motility
nucleic acid binding
RNA binding
mRNA binding
mRNA 3'-UTR binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-hwq2n:80/100.66.77.213:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0