This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Dr03095551_m1
See other POLR2D GE Assays ›
Gene Symbol:
polr2d
Gene Name:
RNA polymerase II subunit D
Gene Aliases:
rnap, wu:fa95b11, zgc:110435
Chromosome Location:
Chr.2: 1124492 - 1129721 on Build GRCz11
Species:
Zebrafish
Species Specific ID (Flybase ID):
-
Assay ID Dr03095551_m1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price 435.00
Your Price
Online offer: 408.65
408.65
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: polr2d
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001002317.2 NP_001002317.2 1 - 2 185 68
GenBank mRNA AY648795.1 - 1 - 2 165 68
BC095261.1 - 1 - 2 79 68
BC164278.1 - 1 - 2 110 68
EH541343.1 - 1 - 2 225 68
EH555518.1 - 1 - 2 222 68

Back To Top

More Information


Set Membership:

Amplicon length less than or equal to 70 Mammalian Gene Collection Recommended Assay Danio rerio Probe spans exons

Panther Classification:

Molecular Function -

DNA-directed RNA polymerase

Gene Ontology Categories:

Function(s) Process(es)

nuclear-transcribed mRNA catabolic process, deadenylation-dependent decay
DNA-templated transcription, initiation
transcription initiation from RNA polymerase II promoter
mRNA export from nucleus in response to heat stress
recruitment of 3'-end processing factors to RNA polymerase II holoenzyme complex
cellular metabolic process
positive regulation of translational initiation
nucleotide binding
single-stranded DNA binding
single-stranded RNA binding
catalytic activity
DNA-directed RNA polymerase activity
translation initiation factor binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-nwsk8:80/100.66.76.192:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0