This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Dr03096140_s1
See other GJA5A GE Assays ›
Gene Symbol:
gja5a
Gene Name:
gap junction protein, alpha 5a
Gene Aliases:
cx45.6, zgc:195347, zgc:195362
Chromosome Location:
Chr.9: 30464402 - 30477594 on Build GRCz11
Species:
Zebrafish
Species Specific ID (Flybase ID):
-
Assay ID Dr03096140_s1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: gja5a
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001007213.1 NP_001007214.1 1 1153 75
XM_005167687.3 XP_005167744.1 2 75
XM_021478635.1 XP_021334310.1 2 75
GenBank mRNA BC109423.1 - 2 1441 75
BC163595.1 - 1 1184 75
BC163596.1 - 1 1184 75
GU363503.1 - 2 1224 75
GU363504.1 - 2 1221 75
GU363505.1 - 2 1464 75
GU363506.1 - 2 1461 75
GU363507.1 - 3 1399 75
GU363508.1 - 3 1396 75

Back To Top

More Information


Set Membership:

Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Detect Genomic DNA Recommended Assay Danio rerio Both primers and probe map within a single exon

Panther Classification:

Molecular Function -

gap junction

Gene Ontology Categories:

Function(s) Process(es)

cell communication
cell-cell signaling
gap junction channel activity

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-blue-7c59b9b899-mnfm6:80/100.66.78.37:80.
git-commit: 8ef7cf686e3c81d28b13492708ce56d167b2f995
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.49.0-Offline