This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Dr03096189_g1
See other CETP GE Assays ›
Gene Symbol:
cetp
Gene Name:
cholesteryl ester transfer protein, plasma
Gene Aliases:
zgc:103675
Chromosome Location:
Chr.18: 17611476 - 17621850 on Build GRCz11
Species:
Zebrafish
Species Specific ID (Flybase ID):
-
Assay ID Dr03096189_g1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: cetp
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001007360.1 NP_001007361.1 7 - 8 688 70
XM_009293552.3 XP_009291827.1 6 - 7 911 70
GenBank mRNA BC085584.1 - 7 - 8 688 70

Back To Top

More Information


Set Membership:

Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Danio rerio Probe spans exons

Gene Ontology Categories:

Function(s) Process(es)

lipid metabolic process
triglyceride metabolic process
lipid transport
steroid metabolic process
cholesterol metabolic process
triglyceride transport
very-low-density lipoprotein particle remodeling
low-density lipoprotein particle remodeling
high-density lipoprotein particle remodeling
cholesterol homeostasis
reverse cholesterol transport
phosphatidylcholine metabolic process
phospholipid homeostasis
triglyceride homeostasis
phospholipid transporter activity
lipid binding
cholesterol binding
triglyceride binding
phosphatidylcholine binding
intermembrane cholesterol transfer activity

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-8rxhs:80/100.66.78.116:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0