This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Dr03098916_m1
See other NAXD GE Assays ›
Gene Symbol:
naxd
Gene Name:
NAD(P)HX dehydratase
Gene Aliases:
carkd, zgc:171429
Chromosome Location:
Chr.9: 8897881 - 8923471 on Build GRCz11
Species:
Zebrafish
Species Specific ID (Flybase ID):
-
Assay ID Dr03098916_m1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: naxd
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001110120.1 NP_001103590.1 10 - 11 1303 62
NM_001328150.1 NP_001315079.1 10 - 11 2293 62
XM_005167456.4 XP_005167513.1 10 - 11 2272 62
XM_005167457.4 XP_005167514.1 10 - 11 2254 62
XM_005167459.2 XP_005167516.1 10 - 11 1361 62
XM_005167460.4 XP_005167517.1 10 - 11 1639 62
XM_005167462.4 XP_005167519.1 10 - 11 1606 62
XM_005167463.4 XP_005167520.1 9 - 10 1322 62
XM_005167464.4 XP_005167521.1 9 - 10 1177 62
GenBank mRNA BC153915.1 - 10 - 11 1303 62

Back To Top

More Information


Set Membership:

Amplicon length less than or equal to 70 3' Most Mammalian Gene Collection Recommended Assay Danio rerio Probe spans exons

Panther Classification:

Molecular Function -

dehydratase

Gene Ontology Categories:

Function(s) Process(es)

nicotinamide nucleotide metabolic process
nucleotide binding
ATP binding
lyase activity
ATP-dependent NAD(P)H-hydrate dehydratase activity
ADP-dependent NAD(P)H-hydrate dehydratase activity

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-2xklz:80/100.66.76.139:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0