This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Dr03099861_m1
See other DHRS9 GE Assays ›
Gene Symbol:
dhrs9
Gene Name:
dehydrogenase/reductase (SDR family) member 9
Gene Aliases:
RDHA, rdh1l, wu:fb64b07, zgc:73286
Chromosome Location:
Chr.9: 48706039 - 48736182 on Build GRCz11
Species:
Zebrafish
Species Specific ID (Flybase ID):
-
Assay ID Dr03099861_m1
Size
Availability Made To Order
Catalog # 4351372
Price 435.00
Your Price
Online offer:
435.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: dhrs9
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_199609.1 NP_955903.1 3 - 4 660 93
GenBank mRNA BC059614.1 - 3 - 4 660 93

Back To Top

More Information


Set Membership:

Amplicon length between 86 and 100 Mammalian Gene Collection Danio rerio Probe spans exons

Panther Classification:

Molecular Function -

dehydrogenase oxidoreductase metabolite interconversion enzyme

Gene Ontology Categories:

Function(s) Process(es)

heart looping
neural crest cell development
exocrine pancreas development
pectoral fin morphogenesis
retinol metabolic process
retinal metabolic process
digestive tract morphogenesis
embryonic viscerocranium morphogenesis
heart contraction
retinol dehydrogenase activity
oxidoreductase activity

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-blue-7c59b9b899-49tpj:80/100.66.77.222:80.
git-commit: 8ef7cf686e3c81d28b13492708ce56d167b2f995
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.49.0-Offline