This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Dr03099945_m1
See other HSD3B7 GE Assays ›
Gene Symbol:
hsd3b7
Gene Name:
hydroxy-delta-5-steroid dehydrogenase, 3 beta- and steroid delta-isomerase
Gene Aliases:
fc37e10, wu:fc37e10, wu:fi12g10, zgc:55799
Chromosome Location:
Chr.3: 239944 - 259334 on Build GRCz11
Species:
Zebrafish
Species Specific ID (Flybase ID):
-
Assay ID Dr03099945_m1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price 435.00
Your Price
Online offer: 408.65
408.65
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: hsd3b7
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_199809.1 NP_956103.1 5 - 6 775 59
XM_009299518.3 XP_009297793.1 8 - 9 1085 59
XM_009299519.3 XP_009297794.1 7 - 8 963 59
XM_017353437.2 XP_017208926.1 6 - 7 1315 59
GenBank mRNA BC045457.1 - 5 - 6 775 59
BC066742.1 - 5 - 6 765 59
BC152680.1 - 6 - 7 1008 59
BC155181.1 - 5 - 6 770 59

Back To Top

More Information


Set Membership:

Amplicon length less than or equal to 70 3' Most Mammalian Gene Collection Recommended Assay Danio rerio Probe spans exons

Panther Classification:

Molecular Function -

dehydratase

Gene Ontology Categories:

Function(s) Process(es)

steroid biosynthetic process
oxidation-reduction process
3-beta-hydroxy-delta5-steroid dehydrogenase activity
oxidoreductase activity
oxidoreductase activity, acting on the CH-OH group of donors, NAD or NADP as acceptor
isomerase activity

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-st8h5:80/100.66.79.144:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0