Assay Details
| Gene Symbol: ergic2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_200407.1 | NP_956701.1 | 8 - 9 | 737 | 91 | |
| GenBank mRNA | BC054593.1 | - | 8 - 9 | 737 | 91 | |
| BC164472.1 | - | 7 - 8 | 607 | 91 | ||
More Information
Set Membership: |
|

ergic2
Genomic Map