This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Dr03100468_m1
See other ROMO1 GE Assays ›
Gene Symbol:
romo1
Gene Name:
reactive oxygen species modulator 1
Gene Aliases:
zgc:73345, zgc:77111
Chromosome Location:
Chr.6: 50401585 - 50404775 on Build GRCz11
Species:
Zebrafish
Species Specific ID (Flybase ID):
-
Assay ID Dr03100468_m1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price 435.00
Your Price
Online offer: 408.65
408.65
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: romo1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_200797.4 NP_957091.1 2 - 3 293 86
GenBank mRNA BC059661.1 - 2 - 3 218 86
BC066639.1 - 2 - 3 203 86
EH450114.1 - 2 - 3 309 86
EH509025.1 - 2 - 3 262 86
EH545431.1 - 2 - 3 316 86

Back To Top

More Information


Set Membership:

Amplicon length between 86 and 100 3' Most Mammalian Gene Collection Recommended Assay Danio rerio Probe spans exons

Gene Ontology Categories:

Function(s)

replicative cell aging
positive regulation of cell proliferation
protein import into mitochondrial matrix
cellular response to reactive oxygen species
defense response to bacterium
protein import into mitochondrial inner membrane
positive regulation of reactive oxygen species metabolic process

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-l4hk6:80/100.66.76.55:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0